Assay Details
| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050214.1 | XP_017905703.1 | 1 - 2 | 3448 | 69 | |
| GenBank mRNA | JT620021.1 | - | 4 - 5 | 1452 | 69 | |
More Information
Set Membership: |
|

PCDH12
Genomic Map