Assay Details
| Gene Symbol: EGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055038.1 | XP_017910527.1 | 2 | 1026 | 75 | |
| XM_018055039.1 | XP_017910528.1 | 2 | 1009 | 75 | ||
More Information
Set Membership: |
|

EGR4
Genomic Map