Assay Details
| Gene Symbol: PTRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967923.2 | XP_013823377.2 | 3 - 4 | 445 | 89 | |
| GenBank mRNA | JO583040.1 | - | 2 - 3 | 560 | 89 | |
More Information
Set Membership: |
|

PTRH1
Genomic Map