Assay Details
| Gene Symbol: LOC106503109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063310.1 | XP_017918799.1 | 8 - 9 | 3823 | 89 | |
| XM_018063311.1 | XP_017918800.1 | 8 - 9 | 3823 | 89 | ||
| XM_018063312.1 | XP_017918801.1 | 9 - 10 | 3448 | 89 | ||
More Information
Set Membership: |
|

LOC106503109
Genomic Map