Assay Details
| Gene Symbol: FUOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041721.1 | XP_017897210.1 | 4 - 5 | 351 | 67 | |
| XM_018041722.1 | XP_017897211.1 | 3 - 4 | 282 | 67 | ||
More Information
Set Membership: |
|

FUOM
Genomic Map