Assay Details
| Gene Symbol: LOC102189713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048912.1 | XP_017904401.1 | 4 - 5 | 595 | 75 | |
| XM_018048919.1 | XP_017904408.1 | 3 - 4 | 419 | 75 | ||
More Information
Set Membership: |
|

LOC102189713
Genomic Map