Assay Details
| Gene Symbol: RXRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677098.3 | XP_005677155.1 | 6 - 7 | 1357 | 57 | |
| XM_005677099.2 | XP_005677156.1 | 5 - 6 | 722 | 57 | ||
| XM_005677100.2 | XP_005677157.1 | 5 - 6 | 633 | 57 | ||
| XM_018046432.1 | XP_017901921.1 | 6 - 7 | 1274 | 57 | ||
| GenBank mRNA | JT754631.1 | - | 7 - 8 | 1089 | 57 | |
More Information
Set Membership: |
|

RXRG
Genomic Map