Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678349.3 | XP_005678406.2 | 6 - 7 | 1227 | 63 | |
| XM_018044687.1 | XP_017900176.1 | 6 - 7 | 1239 | 63 | ||
| XM_018044697.1 | XP_017900186.1 | 6 - 7 | 1814 | 63 | ||
| XM_018044702.1 | XP_017900191.1 | 6 - 7 | 1239 | 63 | ||
| XM_018044707.1 | XP_017900196.1 | 6 - 7 | 1042 | 63 | ||
| GenBank mRNA | JT664560.1 | - | 7 - 8 | 1156 | 63 | |
More Information
Set Membership: |
|

OMA1
Genomic Map