Assay Details
| Gene Symbol: LOC102170588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680604.3 | XP_005680661.1 | 2 - 3 | 205 | 64 | |
| GenBank mRNA | KA320776.1 | - | 1 - 2 | 805 | 64 | |
More Information
Set Membership: |
|

LOC102170588
Genomic Map