Assay Details
| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683059.3 | XP_005683116.1 | 1 | 1474 | 115 | |
| GenBank mRNA | JT659789.1 | - | 2 | 680 | 115 | |
| JT759896.1 | - | 4 | 659 | 115 | ||
More Information
Set Membership: |
|

TAF7
Genomic Map