Assay Details
| Gene Symbol: SCML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966452.2 | XP_013821906.2 | 2 - 3 | 601 | 65 | |
| XM_018053101.1 | XP_017908590.1 | 5 - 6 | 1035 | 65 | ||
More Information
Set Membership: |
|

SCML4
Genomic Map