Assay Details
| Gene Symbol: SRSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054029.1 | XP_017909518.1 | 6 - 7 | 736 | 70 | |
| XM_018054030.1 | XP_017909519.1 | 6 - 7 | 616 | 70 | ||
| XM_018054031.1 | XP_017909520.1 | 6 - 7 | 1103 | 70 | ||
| XM_018054032.1 | XP_017909521.1 | 6 - 7 | 612 | 70 | ||
| XM_018054033.1 | XP_017909522.1 | 6 - 7 | 740 | 70 | ||
| XM_018054034.1 | XP_017909523.1 | 6 - 7 | 615 | 70 | ||
| GenBank mRNA | JO344673.2 | - | 6 - 7 | 745 | 70 | |
More Information
Set Membership: |
|

SRSF5
Genomic Map