Assay Details
| Gene Symbol: GPR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056584.1 | XP_017912073.1 | 2 | 328 | 111 | |
| GenBank mRNA | JO627769.2 | - | 1 | 1057 | 111 | |
More Information
Set Membership: |
|

GPR12
Genomic Map