Assay Details
| Gene Symbol: FZD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058332.1 | XP_017913821.1 | 4 - 5 | 1675 | 63 | |
| XM_018058333.1 | XP_017913822.1 | 4 - 5 | 1700 | 63 | ||
More Information
Set Membership: |
|

FZD6
Genomic Map