Assay Details
| Gene Symbol: RITA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691477.3 | XP_005691534.2 | 3 - 4 | 894 | 68 | |
| XM_005691478.2 | XP_005691535.2 | 3 - 4 | 831 | 68 | ||
| XM_005691480.3 | XP_005691537.2 | 3 - 4 | 894 | 68 | ||
| GenBank mRNA | JO638592.2 | - | 3 - 4 | 730 | 68 | |
More Information
Set Membership: |
|

RITA1
Genomic Map