Assay Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693592.3 | XP_005693649.2 | 2 - 3 | 816 | 64 | |
| GenBank mRNA | KA351406.1 | - | 2 - 3 | 625 | 64 | |
More Information
Set Membership: |
|

TRIM16
Genomic Map