Assay Details
| Gene Symbol: HASPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694161.3 | XP_005694218.2 | 1 | 2433 | 105 | |
| GenBank mRNA | JO614301.2 | - | 1 | 2115 | 105 | |
More Information
Set Membership: |
|

HASPIN
Genomic Map