Assay Details
| Gene Symbol: GABRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694522.3 | XP_005694579.2 | 2 - 3 | 241 | 79 | |
| GenBank mRNA | KA322514.1 | - | 2 - 3 | 284 | 79 | |
More Information
Set Membership: |
|

GABRP
Genomic Map