Assay Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694680.3 | XP_005694737.1 | 4 - 5 | 651 | 103 | |
| XM_005694681.3 | XP_005694738.1 | 4 - 5 | 754 | 103 | ||
| GenBank mRNA | JO637140.2 | - | 1 - 2 | 683 | 103 | |
| JT726439.1 | - | 3 - 4 | 479 | 103 | ||
More Information
Set Membership: |
|

RAB3C
Genomic Map