Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695881.3 | XP_005695938.2 | 3 - 4 | 636 | 103 | |
| XM_018066936.1 | XP_017922425.1 | 3 - 4 | 1866 | 103 | ||
| GenBank mRNA | JP351349.2 | - | 2 - 3 | 789 | 103 | |
More Information
Set Membership: |
|

GLYCTK
Genomic Map