Assay Details
| Gene Symbol: TREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696300.3 | XP_005696357.1 | 1 - 2 | 133 | 76 | |
| GenBank mRNA | JO639225.2 | - | 1 - 2 | 118 | 76 | |
More Information
Set Membership: |
|

TREM2
Genomic Map