Assay Details
| Gene Symbol: LOC102188814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696612.3 | XP_005696669.2 | 2 - 3 | 735 | 82 | |
| XM_013973994.2 | XP_013829448.2 | 2 - 3 | 549 | 82 | ||
More Information
Set Membership: |
|

LOC102188814
Genomic Map