Assay Details
| Gene Symbol: LOC102188814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696612.3 | XP_005696669.2 | 3 - 4 | 1019 | 76 | |
| XM_013973994.2 | XP_013829448.2 | 3 - 4 | 833 | 76 | ||
More Information
Set Membership: |
|

LOC102188814
Genomic Map