Assay Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696985.3 | XP_005697042.2 | 3 - 4 | 1039 | 68 | |
| XM_005696986.3 | XP_005697043.2 | 3 - 4 | 790 | 68 | ||
| GenBank mRNA | JO628909.2 | - | 1 - 2 | 561 | 68 | |
| KA305913.1 | - | 1 - 2 | 341 | 68 | ||
More Information
Set Membership: |
|

ZNF24
Genomic Map