Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697667.3 | XP_005697724.2 | 8 - 9 | 1155 | 55 | |
| XM_005697668.3 | XP_005697725.2 | 8 - 9 | 729 | 55 | ||
More Information
Set Membership: |
|

GDPD3
Genomic Map