Assay Details
| Gene Symbol: SFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698434.3 | XP_005698491.1 | 2 - 3 | 221 | 79 | |
| XM_005698436.3 | XP_005698493.1 | 2 - 3 | 223 | 79 | ||
| XM_018041679.1 | XP_017897168.1 | 2 - 3 | 201 | 79 | ||
More Information
Set Membership: |
|

SFR1
Genomic Map