Assay Details
| Gene Symbol: RAP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044225.1 | XP_017899714.1 | 2 - 3 | 1018 | 89 | |
| XM_018044226.1 | XP_017899715.1 | 2 - 3 | 902 | 89 | ||
| XM_018044227.1 | XP_017899716.1 | 2 - 3 | 1468 | 89 | ||
| GenBank mRNA | JO411037.2 | - | 2 - 3 | 584 | 89 | |
More Information
Set Membership: |
|

RAP2C
Genomic Map