Assay Details
| Gene Symbol: DDX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709665.3 | XP_005709722.3 | 1 | 1625 | 96 | |
| GenBank mRNA | KA318865.1 | - | 1 | 230 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DDX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709665.3 | XP_005709722.3 | 1 | 1625 | 96 | |
| GenBank mRNA | KA318865.1 | - | 1 | 230 | 96 | |
Set Membership: |
|