Assay Details
| Gene Symbol: NR1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041999.1 | XP_017897488.1 | 5 - 6 | 1259 | 73 | |
| GenBank mRNA | JO636575.2 | - | 2 - 3 | 294 | 73 | |
More Information
Set Membership: |
|

NR1D2
Genomic Map