Assay Details
| Gene Symbol: NFE2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047079.1 | XP_017902568.1 | 3 - 4 | 1113 | 93 | |
| GenBank mRNA | JO599466.2 | - | 1 - 2 | 1653 | 93 | |
More Information
Set Membership: |
|

NFE2L3
Genomic Map