Assay Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047956.1 | XP_017903445.1 | 6 - 7 | 784 | 83 | |
| GenBank mRNA | JT688851.1 | - | 2 - 3 | 542 | 83 | |
More Information
Set Membership: |
|

YEATS4
Genomic Map