Assay Details
| Gene Symbol: MAPK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048951.1 | XP_017904440.1 | 9 - 10 | 1089 | 62 | |
| XM_018048952.1 | XP_017904441.1 | 8 - 9 | 732 | 62 | ||
| XM_018048954.1 | XP_017904443.1 | 9 - 10 | 1084 | 62 | ||
| XM_018048955.1 | XP_017904444.1 | 8 - 9 | 737 | 62 | ||
| GenBank mRNA | JO622582.2 | - | 8 - 9 | 765 | 62 | |
| KA329166.1 | - | 1 - 2 | 55 | 62 | ||
More Information
Set Membership: |
|

MAPK11
Genomic Map