Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060680.1 | XP_017916169.1 | 28 - 29 | 3859 | 82 | |
| XM_018060681.1 | XP_017916170.1 | 27 - 28 | 3802 | 82 | ||
| XM_018060682.1 | XP_017916171.1 | 29 - 30 | 5163 | 82 | ||
| XM_018060683.1 | XP_017916172.1 | 29 - 30 | 5163 | 82 | ||
| XM_018060684.1 | XP_017916173.1 | 28 - 29 | 5106 | 82 | ||
More Information
Set Membership: |
|

LOC102184834
Genomic Map