Assay Details
| Gene Symbol: LOC102184834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060681.1 | XP_017916170.1 | 36 - 37 | 4941 | 87 | |
| XM_018060682.1 | XP_017916171.1 | 38 - 39 | 6302 | 87 | ||
| XM_018060683.1 | XP_017916172.1 | 38 - 39 | 6302 | 87 | ||
| XM_018060684.1 | XP_017916173.1 | 37 - 38 | 6245 | 87 | ||
More Information
Set Membership: |
|

LOC102184834
Genomic Map