Assay Details
| Gene Symbol: LOC102188791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044813.1 | XP_017900302.1 | 3 - 4 | 296 | 54 | |
| XM_018044814.1 | XP_017900303.1 | 3 - 4 | 294 | 54 | ||
| XM_018044815.1 | XP_017900304.1 | 3 - 4 | 205 | 54 | ||
| GenBank mRNA | JO610862.2 | - | 3 - 4 | 305 | 54 | |
| JO610863.2 | - | 3 - 4 | 305 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018044815.1 transcripts.
Set Membership: |
|

LOC102188791
Genomic Map