Assay Details
| Gene Symbol: LOC102188791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044813.1 | XP_017900302.1 | 5 - 6 | 481 | 105 | |
| XM_018044814.1 | XP_017900303.1 | 5 - 6 | 479 | 105 | ||
| GenBank mRNA | JO610862.2 | - | 5 - 6 | 490 | 105 | |
More Information
Set Membership: |
|

LOC102188791
Genomic Map