Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064256.1 | XP_017919745.1 | 4 - 5 | 790 | 63 | |
| XM_018064257.1 | XP_017919746.1 | 4 - 5 | 772 | 63 | ||
| GenBank mRNA | KA327263.1 | - | 2 - 3 | 124 | 63 | |
More Information
Set Membership: |
|

RNF135
Genomic Map