Assay Details
| Gene Symbol: LOC102191467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067014.1 | XP_017922503.1 | 2 - 3 | 294 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102191467| Gene Symbol: LOC102191467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067014.1 | XP_017922503.1 | 2 - 3 | 294 | 99 | |
Set Membership: |
|