Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038779.1 | XP_017894268.1 | 9 - 10 | 1251 | 65 | |
| XM_018038780.1 | XP_017894269.1 | 9 - 10 | 1153 | 65 | ||
| XM_018038781.1 | XP_017894270.1 | 9 - 10 | 1251 | 65 | ||
| XM_018038782.1 | XP_017894271.1 | 9 - 10 | 1153 | 65 | ||
| GenBank mRNA | JT750982.1 | - | 9 - 10 | 1187 | 65 | |
More Information
Set Membership: |
|

ZNF76
Genomic Map