Assay Details
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040363.1 | XP_017895852.1 | 3 - 4 | 511 | 66 | |
XM_018040364.1 | XP_017895853.1 | 3 - 4 | 509 | 66 | ||
XM_018040365.1 | XP_017895854.1 | 3 - 4 | 496 | 66 | ||
XM_018040366.1 | XP_017895855.1 | 3 - 4 | 494 | 66 | ||
GenBank mRNA | JO576466.1 | - | 3 - 4 | 569 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
