Assay Details
| Gene Symbol: PNLIPRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975218.2 | XP_013830672.2 | 4 - 5 | 605 | 83 | |
| GenBank mRNA | KA306178.1 | - | 2 - 3 | 219 | 83 | |
More Information
Set Membership: |
|

PNLIPRP3
Genomic Map