Assay Details
| Gene Symbol: SYNPO2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042200.1 | XP_017897689.1 | 1 - 2 | 307 | 54 | |
| GenBank mRNA | JO344206.2 | - | 1 - 2 | 303 | 54 | |
More Information
Set Membership: |
|

SYNPO2L
Genomic Map