Assay Details
| Gene Symbol: SPOCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042489.1 | XP_017897978.1 | 4 - 5 | 830 | 72 | |
| XM_018042490.1 | XP_017897979.1 | 3 - 4 | 657 | 72 | ||
| GenBank mRNA | JO581016.1 | - | 7 - 8 | 1356 | 72 | |
More Information
Set Membership: |
|

SPOCK2
Genomic Map