Assay Details
| Gene Symbol: GSK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674982.3 | XP_005675039.2 | 1 - 2 | 546 | 59 | |
| XM_018042977.1 | XP_017898466.1 | 1 - 2 | 1091 | 59 | ||
| XM_018043043.1 | XP_017898532.1 | 1 - 2 | 548 | 59 | ||
| XM_018043122.1 | XP_017898611.1 | 1 - 2 | 548 | 59 | ||
| XM_018043204.1 | XP_017898693.1 | 1 - 2 | 548 | 59 | ||
| GenBank mRNA | JO343231.2 | - | 10 - 11 | 1266 | 59 | |
| JO346622.2 | - | 1 - 2 | 392 | 59 | ||
| JO346623.2 | - | 1 - 2 | 392 | 59 | ||
| JO391138.2 | - | 9 - 10 | 1034 | 59 | ||
| JO398289.2 | - | 10 - 11 | 1275 | 59 | ||
| JO434752.2 | - | 1 - 2 | 225 | 59 | ||
| JO498713.2 | - | 10 - 11 | 1289 | 59 | ||
| JO561315.2 | - | 1 - 2 | 210 | 59 | ||
| JO585769.2 | - | 11 - 12 | 1326 | 59 | ||
| JO585770.2 | - | 10 - 11 | 1273 | 59 | ||
| JO637745.2 | - | 1 - 2 | 381 | 59 | ||
| JO637746.2 | - | 1 - 2 | 381 | 59 | ||
| KA348397.1 | - | 1 - 2 | 452 | 59 | ||
| KJ649149.1 | - | 1 - 2 | 85 | 59 | ||
| KJ649150.1 | - | 1 - 2 | 85 | 59 | ||
| KJ649151.1 | - | 1 - 2 | 85 | 59 | ||
| KJ649152.1 | - | 1 - 2 | 85 | 59 | ||
| KJ649153.1 | - | 1 - 2 | 85 | 59 | ||
| KJ649154.1 | - | 1 - 2 | 85 | 59 | ||
More Information
Set Membership: |
|

GSK3B
Genomic Map