Assay Details
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676099.3 | XP_005676156.1 | 3 - 4 | 272 | 67 | |
GenBank mRNA | KA323934.1 | - | 3 - 4 | 282 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
