Assay Details
| Gene Symbol: EIF4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676676.2 | XP_005676733.1 | 5 - 6 | 795 | 66 | |
| XM_005676677.3 | XP_005676734.1 | 5 - 6 | 795 | 66 | ||
| XM_005676678.3 | XP_005676735.1 | 5 - 6 | 795 | 66 | ||
| GenBank mRNA | JO644216.2 | - | 5 - 6 | 614 | 66 | |
More Information
Set Membership: |
|

EIF4E2
Genomic Map