Assay Details
| Gene Symbol: DNAJB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046635.1 | XP_017902124.1 | 4 - 5 | 416 | 83 | |
| XM_018046636.1 | XP_017902125.1 | 4 - 5 | 416 | 83 | ||
| XM_018046637.1 | XP_017902126.1 | 4 - 5 | 416 | 83 | ||
| GenBank mRNA | JT624377.1 | - | 3 - 4 | 266 | 83 | |
More Information
Set Membership: |
|

DNAJB6
Genomic Map