Assay Details
| Gene Symbol: AAAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679907.2 | XP_005679964.1 | 5 - 6 | 578 | 59 | |
| GenBank mRNA | JO600552.2 | - | 11 - 12 | 1308 | 59 | |
| JO600555.2 | - | 10 - 11 | 1456 | 59 | ||
More Information
Set Membership: |
|

AAAS
Genomic Map