Assay Details
| Gene Symbol: RASSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047988.1 | XP_017903477.1 | 3 - 4 | 585 | 71 | |
| GenBank mRNA | JO580012.1 | - | 2231 | 71 | ||
More Information
Set Membership: |
|

RASSF3
Genomic Map