Assay Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691406.3 | XP_005691463.2 | 6 - 7 | 427 | 60 | |
| GenBank mRNA | JO428463.1 | - | 8 - 9 | 1993 | 60 | |
More Information
Set Membership: |
|

HPD
Genomic Map